Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011971.4 | NP_036101.1 | 5, 6 | 621 | |
GenBank mRNA | AF060092.1 | - | 5, 6 | 583 | |
AK002331.1 | - | 5, 6 | 622 | ||
AK010488.1 | - | 5, 6 | 602 | ||
AK153162.1 | - | 5, 6 | 654 | ||
BC014783.1 | - | 5, 6 | 588 | 3500033 | |
BC098233.1 | - | 4, 5 | 583 | 1547611 |