Gene Transcripts

Gene Symbol
Ankdd1b
Entrez Gene ID
271144
Gene Name
ankyrin repeat and death domain containing 1B
Gene Aliases
9330128J19Rik
Chromosome Location
Chr. 13: 96415030 - 96471354 on Build GRCm38
UniGene ID
Mm.470057
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042714.1 NP_001036179.1 2 264
GenBank mRNA BC112417.1 - 2 264

View Full Product Details