Gene Transcripts
- Gene Symbol
- Dok3
- Entrez Gene ID
- 27261
- Gene Name
- docking protein 3
- Gene Aliases
- AI450713, Dokl
- Chromosome Location
- Chr. 13: 55523235 - 55528538 on Build GRCm38
- UniGene ID
- Mm.33910
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013739.2 |
NP_038767.1
|
3, 4 |
501 |
|
GenBank mRNA |
AF179242.1 |
- |
3, 4 |
501 |
|
AF237580.1 |
- |
3, 4 |
497 |
|
AK137520.1 |
- |
3, 4 |
488 |
|
AK154412.1 |
- |
2, 3 |
1324 |
|
AK154750.1 |
- |
2, 3 |
1324 |
|
AK155180.1 |
- |
2, 3 |
1331 |
|
AK156060.1 |
- |
2, 3 |
1331 |
|
AK172511.1 |
- |
2, 3 |
1331 |
|
BC048784.1 |
- |
2, 3 |
1632 |
|
BC116922.1 |
- |
3, 4 |
500 |
40126241 |
BC120535.1 |
- |
3, 4 |
500 |
40129458 |
View Full Product Details