Gene Transcripts

Gene Symbol
Mgea5
Entrez Gene ID
76055
Gene Name
meningioma expressed antigen 5 (hyaluronidase)
Gene Aliases
2810009A20Rik, 4833427O07Rik, 5830447M11Rik, AA408215, Hy5, Ncoat, OGA, mKIAA0679
Chromosome Location
Chr. 19: 45750255 - 45783832 on Build GRCm38
UniGene ID
Mm.122725
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK045615.1 - 1 533
AK054352.1 - 1 543
BC147259.1 - 1 132
BC147260.1 - 1 132

View Full Product Details