Gene Transcripts

Gene Symbol
Fnd3c2
Entrez Gene ID
331491
Gene Name
fibronectin type III domain containing 3C2
Gene Aliases
5031408O05Rik, Gm775
Chromosome Location
Chr. X: 106235246 - 106255372 on Build GRCm38
UniGene ID
Mm.426150
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033424.3 NP_001028596.2 2 555
GenBank mRNA AK030291.1 - 2 559
AK146031.1 - 2 556

View Full Product Details