Gene Transcripts

Gene Symbol
Pigg
Entrez Gene ID
433931
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class G
Gene Aliases
Gpi7
Chromosome Location
Chr. 5: 108312783 - 108349359 on Build GRCm38
UniGene ID
Mm.319881
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081234.1 NP_001074703.1 11 2505
GenBank mRNA BC051059.1 - 5 1386

View Full Product Details