Gene Transcripts

Gene Symbol
Mink1
Entrez Gene ID
50932
Gene Name
misshapen-like kinase 1 (zebrafish)
Gene Aliases
B55, MINK, Map4k6, Ysk2
Chromosome Location
Chr. 11: 70562594 - 70614482 on Build GRCm38
UniGene ID
Mm.42967
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001045959.1 NP_001039424.1 22, 23 2869
NM_001045964.1 NP_001039429.1 23, 24 2890
NM_016713.2 NP_057922.2 21, 22 2758
NM_176893.2 NP_795712.2 22, 23 2782
GenBank mRNA AB035697.1 - 21, 22 2743
AB041925.1 - 22, 23 2768
AK220178.1 - 4, 5 308
BC011346.1 - 18, 19 2262
BC052474.1 - 22, 23 2724 6833893
BC079549.1 - 21, 22 3240
BU707794.1 - 7, 8 599

View Full Product Details