Gene Transcripts
- Gene Symbol
- Ankzf1
- Entrez Gene ID
- 52231
- Gene Name
- ankyrin repeat and zinc finger domain containing 1
- Gene Aliases
- 1300008P06Rik, 2810025E10Rik, D1Ertd161e
- Chromosome Location
- Chr. 1: 75192160 - 75199387 on Build GRCm38
- UniGene ID
- Mm.259326
- Species
- Mus musculus
- Gene Symbol
- Glb1l
- Entrez Gene ID
- 74577
- Gene Name
- galactosidase, beta 1-like
- Gene Aliases
- 4833408P15Rik
- Chromosome Location
- Chr. 1: 75198235 - 75210816 on Build GRCm38
- UniGene ID
- Mm.458319
- Species
- Mus musculus
View Full Product Details