Gene Transcripts

Gene Symbol
Gal3st3
Entrez Gene ID
545276
Gene Name
galactose-3-O-sulfotransferase 3
Gene Aliases
Chromosome Location
Chr. 19: 5298331 - 5308739 on Build GRCm38
UniGene ID
Mm.447174
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024717.2 NP_001019888.1 3 583
GenBank mRNA BC066996.1 - 3 573
BU612752.1 - 3 583

View Full Product Details