Gene Transcripts

Gene Symbol
Ift20
Entrez Gene ID
55978
Gene Name
intraflagellar transport 20
Gene Aliases
0610009H04Rik, AU015496
Chromosome Location
Chr. 11: 78536436 - 78541473 on Build GRCm38
UniGene ID
Mm.358671
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018854.4 NP_061342.1 5, 6 694
GenBank mRNA AK002397.1 - 5, 6 695
AK139942.1 - 5, 6 731
AY082613.1 - 4, 5 414
U38981.1 - 527

View Full Product Details