Gene Transcripts

Gene Symbol
Terf2ip
Entrez Gene ID
57321
Gene Name
telomeric repeat binding factor 2, interacting protein
Gene Aliases
Rap1
Chromosome Location
Chr. 8: 112011359 - 112020528 on Build GRCm38
UniGene ID
Mm.213064
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020584.2 NP_065609.2 1 735
GenBank mRNA AB041557.1 - 1 689
AB041559.1 - 1 690
AK041613.1 - 1 681
AK047222.1 - 1 693
AK051818.1 - 1 655
AK053864.1 - 1 691
AK081557.1 - 1 691
AK084563.1 - 1 683
AK087729.1 - 1 693
AK148508.1 - 1 695
AK148537.1 - 1 697
AK161452.1 - 1 714
AK163044.1 - 1 691
AK165074.1 - 1 693
AK213108.1 - 1 242
BC012270.1 - 1 651 3483521
BC017641.1 - 1 711 4483320

View Full Product Details