Gene Transcripts

Gene Symbol
Shank3
Entrez Gene ID
58234
Gene Name
SH3/ankyrin domain gene 3
Gene Aliases
AI841104, SHANK3c-3, SHANK3c-4, Shank3b
Chromosome Location
Chr. 15: 89499707 - 89560261 on Build GRCm38
UniGene ID
Mm.146855
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021423.3 NP_067398.2 8, 9 961
GenBank mRNA AB231013.1 - 8, 9 961
AJ245904.1 - 6, 7 815
KF612592.1 - 4, 5 477

View Full Product Details