Gene Transcripts

Gene Symbol
Vmn2r59
Entrez Gene ID
628444
Gene Name
vomeronasal 2, receptor 59
Gene Aliases
EG628444
Chromosome Location
Chr. 7: 42011792 - 42058981 on Build GRCm38
UniGene ID
Mm.469901
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105056.1 NP_001098526.1 2 348

View Full Product Details