Gene Transcripts

Gene Symbol
Sult2a7
Entrez Gene ID
638251
Gene Name
sulfotransferase family 2A, dehydroepiandrosterone (DHEA)-preferring, member 7
Gene Aliases
EG638251, Gm7231
Chromosome Location
Chr. 7: 14465159 - 14492926 on Build GRCm38
UniGene ID
Mm.482063
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184981.1 NP_001171910.1 5 542

View Full Product Details