Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025564.2 | NP_079840.2 | 3, 4 | 344 | |
GenBank mRNA | AK006303.1 | - | 3, 4 | 334 | |
AK007769.1 | - | 3, 4 | 331 | ||
AK008200.1 | - | 3, 4 | 346 | ||
AK013310.1 | - | 3, 4 | 329 | ||
BC027505.1 | - | 3, 4 | 317 | 3372083 | |
KF051005.1 | - | 3, 4 | 265 |