Gene Transcripts

Gene Symbol
Ndufaf3
Entrez Gene ID
66706
Gene Name
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 3
Gene Aliases
4733401H18Rik
Chromosome Location
Chr. 9: 108565865 - 108567342 on Build GRCm38
UniGene ID
Mm.29471
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023247.1 NP_075736.1 5 915
GenBank mRNA AF235017.1 - 5 915
AK005252.1 - 5 530
AK014634.1 - 5 530
BC027513.1 - 5 948 5118121

View Full Product Details