Gene Transcripts
- Gene Symbol
- 4933437I04Rik
- Entrez Gene ID
- 71246
- Gene Name
- RIKEN cDNA 4933437I04 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.23173
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK017101.1 |
- |
1 |
74 |
|
- Gene Symbol
- Tomm34
- Entrez Gene ID
- 67145
- Gene Name
- translocase of outer mitochondrial membrane 34
- Gene Aliases
- 2610100K07Rik, TOM34
- Chromosome Location
- Chr. 2: 164053538 - 164071177 on Build GRCm38
- UniGene ID
- Mm.23173
- Species
- Mus musculus
View Full Product Details