Gene Transcripts

Gene Symbol
Coq9
Entrez Gene ID
67914
Gene Name
coenzyme Q9 homolog (yeast)
Gene Aliases
2310005O14Rik, C78387
Chromosome Location
Chr. 8: 94838417 - 94854895 on Build GRCm38
UniGene ID
Mm.169234
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026452.2 NP_080728.1 7 733
GenBank mRNA AK004527.1 - 3 158
AK009179.1 - 7 746
AK158979.1 - 7 752

View Full Product Details