Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026455.4 | NP_080731.1 | 1 | 99 | |
GenBank mRNA | AK007524.1 | - | 1 | 97 | |
AK008285.1 | - | 1 | 78 | ||
AK008336.1 | - | 1 | 85 | ||
AK011922.1 | - | 1 | 85 | ||
AK075976.1 | - | 1 | 100 | ||
AK082792.1 | - | 1 | 70 | ||
AK150416.1 | - | 1 | 70 | ||
BC006058.1 | - | 1 | 51 | 3591395 | |
BC085487.1 | - | 1 | 65 |