Gene Transcripts
- Gene Symbol
- Ly6g6c
- Entrez Gene ID
- 68468
- Gene Name
- lymphocyte antigen 6 complex, locus G6C
- Gene Aliases
- 1110003M04Rik, AU016360, G6c, NG24
- Chromosome Location
- Chr. 17: 35067325 - 35070048 on Build GRCm38
- UniGene ID
- Mm.215096
- Species
- Mus musculus
- Gene Symbol
- Catsper3
- Entrez Gene ID
- 76856
- Gene Name
- cation channel, sperm associated 3
- Gene Aliases
- 4921522D01Rik, CACRC
- Chromosome Location
- Chr. 13: 55784579 - 55824942 on Build GRCm38
- UniGene ID
- Mm.159795
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC089518.1 |
- |
|
244 |
|
View Full Product Details