Gene Transcripts

Gene Symbol
Lyzl4
Entrez Gene ID
69032
Gene Name
lysozyme-like 4
Gene Aliases
1810009N24Rik, AI504271, Lyc4
Chromosome Location
Chr. 9: 121577841 - 121641566 on Build GRCm38
UniGene ID
Mm.33351
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026915.2 NP_081191.1 4, 5 667
GenBank mRNA AK007412.1 - 636
AY572449.1 - 2, 3 318
BC024932.1 - 4, 5 667 3985345

View Full Product Details