Gene Transcripts

Gene Symbol
Ankrd60
Entrez Gene ID
70065
Gene Name
ankyrin repeat domain 60
Gene Aliases
1700019A24Rik, 1700030G11Rik
Chromosome Location
Chr. 2: 173568661 - 173578341 on Build GRCm38
UniGene ID
Mm.425115
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027303.1 NP_081579.1 4 811
GenBank mRNA AK006546.1 - 4 754

View Full Product Details