Gene Transcripts

Gene Symbol
Ccdc81
Entrez Gene ID
70884
Gene Name
coiled-coil domain containing 81
Gene Aliases
4921513D09Rik
Chromosome Location
Chr. 7: 89866148 - 89903629 on Build GRCm38
UniGene ID
Mm.252823
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162979.1 NP_001156451.1 14, 15 2202
GenBank mRNA AK014876.1 - 14, 15 2185
AK029544.1 - 15, 16 2300
AV255663.2 - 4, 5 502

View Full Product Details