Gene Transcripts

Gene Symbol
Nipbl
Entrez Gene ID
71175
Gene Name
Nipped-B homolog (Drosophila)
Gene Aliases
Idn3
Chromosome Location
Chr. 15: 8289824 - 8465310 on Build GRCm38
UniGene ID
Mm.240329
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027707.3 NP_081983.2 36, 37 6740
NM_201232.2 NP_957684.1 36, 37 6740
GenBank mRNA AJ627033.1 - 36, 37 6740
AJ640138.1 - 36, 37 6740
AK135347.1 - 8, 9 787
AK164261.1 - 3, 4 263

View Full Product Details