Gene Transcripts

Gene Symbol
Cyp2u1
Entrez Gene ID
71519
Gene Name
cytochrome P450, family 2, subfamily u, polypeptide 1
Gene Aliases
8430436A10Rik
Chromosome Location
Chr. 3: 131290491 - 131303227 on Build GRCm38
UniGene ID
Mm.473179
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027816.3 NP_082092.2 1, 2 554
GenBank mRNA AK018458.1 - 1, 2 530
AK041477.1 - 1, 2 555
AK142740.1 - 1, 2 494

View Full Product Details