Gene Transcripts

Gene Symbol
Fezf1
Entrez Gene ID
73191
Gene Name
Fez family zinc finger 1
Gene Aliases
3110069A13Rik, Fez, Zfp312-like, fez-like
Chromosome Location
Chr. 6: 23245044 - 23255368 on Build GRCm38
UniGene ID
Mm.55115
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028462.1 NP_082738.1 3 1170
GenBank mRNA AK014242.1 - 3 1048
BC049157.1 - 3 1269

View Full Product Details