Gene Transcripts

Gene Symbol
Gsdmc4
Entrez Gene ID
74548
Gene Name
gasdermin C4
Gene Aliases
9030605I04Rik
Chromosome Location
Chr. 15: 63891257 - 63912316 on Build GRCm38
UniGene ID
Mm.273185
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028992.1 NP_083268.1 6 829
GenBank mRNA AK018537.1 - 6 830
AK163061.1 - 6 830

View Full Product Details