Gene Transcripts

Gene Symbol
Armc4
Entrez Gene ID
74934
Gene Name
armadillo repeat containing 4
Gene Aliases
4930463I21Rik, b2b227.1Clo, b2b643Clo
Chromosome Location
Chr. 18: 7088233 - 7301128 on Build GRCm38
UniGene ID
Mm.107918
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081393.1 NP_001074862.1 14 2045
GenBank mRNA BC158097.1 - 13 2034

View Full Product Details