Gene Transcripts

Gene Symbol
Nop14
Entrez Gene ID
75416
Gene Name
NOP14 nucleolar protein
Gene Aliases
2610033H07Rik, Nol14
Chromosome Location
Chr. 5: 34638536 - 34660148 on Build GRCm38
UniGene ID
Mm.40292
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029278.2 NP_083554.2 1 199
GenBank mRNA AK011648.1 - 1 175
AK078605.1 - 1 184
AK079609.1 - 1 201
AK148605.1 - 1 158
AK161728.1 - 1 137
BC024998.1 - 1 120 5012070
BC043043.1 - 1 142 5701430

View Full Product Details