Gene Transcripts

Gene Symbol
Raph1
Entrez Gene ID
77300
Gene Name
Ras association (RalGDS/AF-6) and pleckstrin homology domains 1
Gene Aliases
9430025M21Rik, C730009O10Rik, Lpd
Chromosome Location
Chr. 1: 60483185 - 60567074 on Build GRCm38
UniGene ID
Mm.440325
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001045513.3 NP_001038978.3 13 1861
GenBank mRNA DV652147.1 - 5 402

View Full Product Details