Gene Transcripts

Gene Symbol
Shisa4
Entrez Gene ID
77552
Gene Name
shisa family member 4
Gene Aliases
9330132O05Rik, Tmem58, mShisa4
Chromosome Location
Chr. 1: 135371057 - 135375277 on Build GRCm38
UniGene ID
Mm.41407
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175259.4 NP_780468.1 4 510
GenBank mRNA AK039457.1 - 4 521
AK134719.1 - 2 293
BC044813.1 - 4 510 4950177

View Full Product Details