Gene Transcripts

Gene Symbol
Nrg4
Entrez Gene ID
83961
Gene Name
neuregulin 4
Gene Aliases
AI552600
Chromosome Location
Chr. 9: 55220222 - 55283625 on Build GRCm38
UniGene ID
Mm.443874
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032002.2 NP_114391.1 5 486
GenBank mRNA AF083067.2 - 4 364
AK041077.1 - 6 518
AK080058.1 - 5 438
AK080161.1 - 7 580
BC034839.1 - 8 779 3372942

View Full Product Details