Gene Transcripts
- Gene Symbol
- Trim7
- Entrez Gene ID
- 94089
- Gene Name
- tripartite motif-containing 7
- Gene Aliases
- AI790312
- Chromosome Location
- Chr. 11: 48826138 - 48850644 on Build GRCm38
- UniGene ID
- Mm.390335
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053166.2 |
NP_444396.2
|
3, 4 |
464 |
|
GenBank mRNA |
AF396656.1 |
- |
3, 4 |
479 |
|
View Full Product Details