Product Details

SNP ID
rs13475987
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:93403982 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TTGCCTCCTGTGTACCTTCCAGTAC[C/T]GGGCCAGAAACTCAGCACACAGGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Ano7 PubMed Links

Gene Details

Gene
Ano7
Gene Name
anoctamin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271884.1 3697 UTR 3 NP_001258813.1
NM_207031.2 3697 UTR 3 NP_996914.1
Gene
Hdlbp
Gene Name
high density lipoprotein (HDL) binding protein
There are no transcripts associated with this gene.

View Full Product Details