Product Details

SNP ID
rs13481473
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.12:57325623 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AAGGAGAGAGACGAAGCCATTGCAC[A/G]TGTGAAACATATGGAAATGTCACTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Mipol1 PubMed Links

Gene Details

Gene
Mipol1
Gene Name
mirror-image polydactyly gene 1 homolog (human)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164370.1 727 Missense Mutation NP_001157842.1

View Full Product Details