Product Details

SNP ID
rs13477042
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.3:30947054 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ATCACACAAAGTAATATCTGCTCAA[C/T]TGTTTAGCTGAAAGTTTACTGAACG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Phc3 PubMed Links

Gene Details

Gene
Phc3
Gene Name
polyhomeotic-like 3 (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165954.1 Intron NP_001159426.1
NM_001165955.1 Intron NP_001159427.1
NM_001165956.1 Intron NP_001159428.1
NM_153421.2 Intron NP_700470.2

View Full Product Details