Product Details

SNP ID
rs6195594
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:143594736 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGATCAAATTTTCTGCAACTAAGAT[A/G]TGACCTATTATTTGTCTTAAATAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Pcsk2 PubMed Links

Gene Details

Gene
Pcsk2
Gene Name
proprotein convertase subtilisin/kexin type 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_008792.4 Intron NP_032818.1

View Full Product Details