Product Details
- SNP ID
-
rs13476796
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:137085033 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGCTGGAGGCTGGAGGACCGACA[C/T]TCGCCAACACCAGTGCAGGGGCGCA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Jag1
PubMed Links
Gene Details
- Gene
- Jag1
- Gene Name
- jagged 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_013822.5 |
3068 |
Silent Mutation |
|
|
NP_038850.1 |
- Gene
- Slx4ip
- Gene Name
- SLX4 interacting protein
There are no transcripts associated with this gene.
View Full Product Details