Product Details

SNP ID
rs3717610
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:5753110 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CATACAAGAATATTTACAGAAAAAT[G/T]AAAAGCTTACAAATGGAAAGACTGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
2210015D19Rik PubMed Links

Gene Details

Gene
2210015D19Rik
Gene Name
RIKEN cDNA 2210015D19 gene
There are no transcripts associated with this gene.

Gene
Urgcp
Gene Name
upregulator of cell proliferation
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077661.1 Intron NP_001071129.1
NM_178623.3 Intron NP_848738.2

View Full Product Details