Product Details

SNP ID
rs13482654
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.15:76694306 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCTGGCTCTGTTCACAGGACGATGG[C/T]CCTGAGGTGGCCAGACCACACAATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Gpt PubMed Links

Gene Details

Gene
Gpt
Gene Name
glutamic pyruvic transaminase, soluble
There are no transcripts associated with this gene.

Gene
Mfsd3
Gene Name
major facilitator superfamily domain containing 3
There are no transcripts associated with this gene.

Gene
Ppp1r16a
Gene Name
protein phosphatase 1, regulatory (inhibitor) subunit 16A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033371.2 2014 Silent Mutation NP_203535.1
Gene
Recql4
Gene Name
RecQ protein-like 4
There are no transcripts associated with this gene.

View Full Product Details