Product Details
- SNP ID
-
rs13476124
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:135584727 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGTTCGTCATCCGATTTGCCATC[A/T]TCGCTTACCGCTGCCTCAGGCGCCT
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Gm4793
PubMed Links
Gene Details
- Gene
- Gm4793
- Gene Name
- predicted gene 4793
There are no transcripts associated with this gene.
- Gene
- Nav1
- Gene Name
- neuron navigator 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_173437.2 |
629 |
Missense Mutation |
|
|
NP_775613.2 |
View Full Product Details