Product Details
- SNP ID
-
rs3716716
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:103008495 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCAATTGAAAGTTGGATTTATAAA[T/G]CCTGTCCAGTTGACTAGACTTTAAA
- Phenotype
-
- Polymorphism
- T/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Alx1
PubMed Links
Gene Details
- Gene
- Alx1
- Gene Name
- ALX homeobox 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_172553.4 |
1845 |
UTR 3 |
|
|
NP_766141.1 |
View Full Product Details