Product Details

SNP ID
rs13483127
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.17:81385824 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCGAGTGTGCCTGTCCCCCTAAAGC[C/T]GTGCACAATACACACAAAATAAAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Slc8a1 PubMed Links

Gene Details

Gene
Slc8a1
Gene Name
solute carrier family 8 (sodium/calcium exchanger), member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001112798.2 5890 UTR 3 NP_001106269.1
NM_001286684.1 5890 UTR 3 NP_001273613.1
NM_011406.3 5890 UTR 3 NP_035536.2

View Full Product Details