Product Details

SNP ID
rs3702010
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:43043786 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTTCTGGGACCCTCCATGTAGTGAT[T/G]CCAAGCACACAGCTGCTCCCACCAG
Phenotype
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
Sobp PubMed Links

Gene Details

Gene
Sobp
Gene Name
sine oculis-binding protein homolog (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175407.3 Intron NP_780616.4

View Full Product Details