Product Details
- SNP ID
-
rs13483139
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:84722786 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATAAGTTTCACTGCTAAATACAAC[A/G]TTTTGCTTTTCGGCTTTAAGGAAGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Lrpprc
PubMed Links
Gene Details
- Gene
- Lrpprc
- Gene Name
- leucine-rich PPR-motif containing
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_028233.2 |
3456 |
Silent Mutation |
|
|
NP_082509.2 |
View Full Product Details