Product Details

SNP ID
rs13476394
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:27132630 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CAGAGCAGAGCGTCGTGCTCAAGGG[C/T]GCACCCCAGCCAGCCAGGGGAATTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Fam163b PubMed Links

Gene Details

Gene
Fam163b
Gene Name
family with sequence similarity 163, member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175427.4 Intron NP_780636.1

View Full Product Details