Product Details
- SNP ID
-
rs13481783
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:42840147 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAAATGCATAGCAGGTAGGACCAA[C/T]GAATATACTTGAAATGGCAATTCAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Phactr1
PubMed Links
Gene Details
- Gene
- Phactr1
- Gene Name
- phosphatase and actin regulator 1
View Full Product Details