Product Details
- SNP ID
-
rs29378324
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:4880068 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGACCGCTAGTTCCTGATTTTGATT[G/A]TATTGAATCAAATCAGATTTGAATG
- Phenotype
-
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Esr1
PubMed Links
Gene Details
- Gene
- Esr1
- Gene Name
- estrogen receptor 1 (alpha)
There are no transcripts associated with this gene.
View Full Product Details