Product Details

SNP ID
rs13481970
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:99353310 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CGGGAAATGTGCGCTCATGTGGAGC[G/T]TCGTGATTGGACATCGCGCTGGGAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Mrps27 PubMed Links

Gene Details

Gene
Mrps27
Gene Name
mitochondrial ribosomal protein S27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173757.3 Intron NP_776118.2
Gene
Ptcd2
Gene Name
pentatricopeptide repeat domain 2
There are no transcripts associated with this gene.

View Full Product Details