Product Details

SNP ID
rs3681808
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.15:13042334 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AAACTTAAGACTGATTTGGAATCCG[C/T]GTCAGCCTTGGAGACTGATGAACTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Cdh6 PubMed Links

Gene Details

Gene
Cdh6
Gene Name
cadherin 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007666.3 Intron NP_031692.2

View Full Product Details